Please login to your online account to display your discounted pricing

Sulfosalicylic Acid, Certified, 5.0% (w/v) ±0.1%, LabChem™

$59.83 - $59.83

Chemical Identifiers

Chemical Name or Material Sulfosalicylic Acid
CAS 7732-18-5
CAS Max % 95
CAS 5965-83-3
CAS Max % 5
Purity Grade Certified Grade
Linear Formula C7H6O6S·2H2O
Formula Weight 254.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC255401

 
lab chem
LC255401
Amber Poly 500mL Each for $59.83
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 254.21
Assay 5.0% ± 0.1% (w/v)
Identification Passes Test
CAS 7732-18-5
CAS 5965-83-3
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Packaging Amber Poly
CAS Max % 95
Solubility Information Soluble in water
Health Hazard 1 Warning
Chemical Name or Material Sulfosalicylic Acid
Physical Form Liquid
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
CAS Max % 5
Color Clear, colorless
Linear Formula C7H6O6S·2H2O
Recommended Storage Room Temperature
Quantity 500mL
Safety and Handling

Warning

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

Room Temperature