Please login to your online account to display your discounted pricing

Sulfosalicylic Acid, Certified, 20.0% (w/v) ±0.5%, LabChem™

$129.63 - $129.63

Chemical Identifiers

Chemical Name or Material Sulfosalicylic Acid
CAS 7732-18-5
CAS Max % 80
CAS 5965-83-3
CAS Max % 20
Purity Grade Certified Grade
Linear Formula C7H6O6S·2H2O
Formula Weight 254.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC255451

 
lab chem
LC255451
Amber Poly 500mL Each for $129.63
Description & Specifications

Specifications

Assay 20.0% ± 0.5% (w/v)
Formula Weight 254.21
Purity Grade Certified Grade
Quantity 500mL
CAS 5965-83-3
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Identification Passes Test
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS Max % 20
Recommended Storage Room Temperature
Packaging Amber Poly
Chemical Name or Material Sulfosalicylic Acid
Color Clear to slight pink
CAS Max % 80
Physical Form Liquid
CAS 7732-18-5
Health Hazard 1 Warning
Linear Formula C7H6O6S·2H2O
Solubility Information Soluble in water
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Room Temperature