Please login to your online account to display your discounted pricing

Sulfosalicylic Acid, 10% (w/v) Aqueous Solution, Ricca Chemical


Quantity 20L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR812500020F

 View more versions of this product

Catalog No. R812500020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 20L
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 93.0
Packaging Cubitainer
CAS 5965-83-3
Concentration or Composition (by Analyte or Components) 10%(w/v)
CAS Min % 8.91
CAS Max % 9.28

Specifications

CAS 7732-18-5
CAS Min % 89.0
CAS Max % 93.0
CAS 5965-83-3
CAS Min % 8.91
CAS Max % 9.28