Please login to your online account to display your discounted pricing

Sulforhodamine B, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC333130050

 View more versions of this product

Catalog No. AC333130050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 558.67
Quantity 5g
Color Dark green
Beilstein 18, II, 491
Chemical Name or Material Sulforhodamine B
Assay Percent Range 97%
MDL Number MFCD00067690
CAS 2609-88-3
Packaging Glass bottle
Molecular Formula C27H30N2O7S2
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Sulforhodamine B
CAS 2609-88-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C27H30N2O7S2
Formula Weight 558.67
MDL Number MFCD00067690