Please login to your online account to display your discounted pricing

Sulforhodamine B, 97%, ACROS Organics™

$125.88 - $125.88

Chemical Identifiers

Chemical Name or Material Sulforhodamine B
CAS 2609-88-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C27H30N2O7S2
Formula Weight 558.67
MDL Number MFCD00067690
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC333130050

  SDS
thermo fisher scientific
AC333130050
Glass bottle 5g Each for $125.88

AC333130010

  SDS
fisher scientific acros
333130010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 558.67
Quantity 5g
Color Dark green
Beilstein 18, II, 491
Chemical Name or Material Sulforhodamine B
Assay Percent Range 97%
MDL Number MFCD00067690
CAS 2609-88-3
Packaging Glass bottle
Molecular Formula C27H30N2O7S2
Infrared Spectrum Authentic

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.