Please login to your online account to display your discounted pricing

Sulforhodamine B, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  333130010

 View more versions of this product

Catalog No. AC333130010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 558.67
Quantity 1g
Color Dark green
Beilstein 18, II, 491
Chemical Name or Material Sulforhodamine B
CAS 2609-88-3
Assay Percent Range 97%
Molecular Formula C27H30N2O7S2
MDL Number MFCD00067690
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material Sulforhodamine B
CAS 2609-88-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C27H30N2O7S2
Formula Weight 558.67
MDL Number MFCD00067690