missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Sulforhodamine 101 (free acid), 99%, pure, laser grade
$191.18 - $191.18
Chemical Identifiers
CAS | 60311-02-6 |
---|---|
Molecular Formula | C31H29ClN2O6S2 |
Molecular Weight (g/mol) | 625.15 |
MDL Number | MFCD00012407 |
InChI Key | MPLHNVLQVRSVEE-UHFFFAOYSA-N |
Synonym | sulforhodamine 101, sulforhodamine 101; sulforhodamine 640, unii-fx0es3271v, texas red, sulforhodamine 101 acid chloride, 2-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-pyrido 3,2,1-ij quinolizino 1',9':6,7,8 chromeno 2,3-f quinolin-4-ium-9-yl-5-sulfobenzenesulfonate, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9$l^ 5 ,23-diazaheptacyclo 17.7.1.1^ 5,9 .0^ 2,17 .0^ 4,15 .0^ 23,27 .0^ 13,28 octacosa-1 27 ,2 17 ,4,9 28 ,13,15,18-heptaen-9-ylium, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9??,23-diazaheptacyclo 17.7.1.1?,?.0 2 , 1 ?.0?, 1 ?.0 2 3 , 2 ?.0 1 3 , 2 ? octacosa-1 27 ,2 17 ,4,9 28 ,13,15,18-heptaen-9-ylium, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9lambda,23-diazaheptacyclo 17.7.1.1,.0 2 , 1 .0, 1 .0 2 3 , 2 .0 1 3 , 2 octacosa-1,4,9 28 ,13,15,17,19 27-heptaen-9-ylium, 9-2,4-disulfophenyl-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-xantheno 2,3,4-ij:5,6,7-i'j' diquinolizin-18-ium inner salt, hydrogen 9-2,4-disulphonatophenyl-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-xantheno 2,3,4-ij:5,6,7-i'j' diquinolizin-18-ium |
PubChem CID | 122180 |
ChEBI | CHEBI:52274 |
IUPAC Name | 16-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3λâ´-oxa-9,23-diazaheptacyclo[17.7.1.1âµ,â¹.0²,¹â·.0â´,¹âµ.0²³,²â·.0¹³,²â¸]octacosa-1,3,5(28),13,15,17,19(27)-heptaen-3-ylium |
SMILES | [O-]S(=O)(=O)C1=CC(=CC=C1C1=C2C=C3CCCN4CCCC(C2=[O+]C2=C5CCCN6CCCC(C=C12)=C56)=C34)S(Cl)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC419941000
|
Thermo Scientific Chemicals
419941000 |
100 mg |
Each for $191.18
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
60311-02-6 | |
625.15 | |
MPLHNVLQVRSVEE-UHFFFAOYSA-N | |
122180 | |
16-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3λâ´-oxa-9,23-diazaheptacyclo[17.7.1.1âµ,â¹.0²,¹â·.0â´,¹âµ.0²³,²â·.0¹³,²â¸]octacosa-1,3,5(28),13,15,17,19(27)-heptaen-3-ylium |
C31H29ClN2O6S2 | |
MFCD00012407 | |
sulforhodamine 101, sulforhodamine 101; sulforhodamine 640, unii-fx0es3271v, texas red, sulforhodamine 101 acid chloride, 2-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-pyrido 3,2,1-ij quinolizino 1',9':6,7,8 chromeno 2,3-f quinolin-4-ium-9-yl-5-sulfobenzenesulfonate, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9$l^ 5 ,23-diazaheptacyclo 17.7.1.1^ 5,9 .0^ 2,17 .0^ 4,15 .0^ 23,27 .0^ 13,28 octacosa-1 27 ,2 17 ,4,9 28 ,13,15,18-heptaen-9-ylium, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9??,23-diazaheptacyclo 17.7.1.1?,?.0 2 , 1 ?.0?, 1 ?.0 2 3 , 2 ?.0 1 3 , 2 ? octacosa-1 27 ,2 17 ,4,9 28 ,13,15,18-heptaen-9-ylium, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9lambda,23-diazaheptacyclo 17.7.1.1,.0 2 , 1 .0, 1 .0 2 3 , 2 .0 1 3 , 2 octacosa-1,4,9 28 ,13,15,17,19 27-heptaen-9-ylium, 9-2,4-disulfophenyl-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-xantheno 2,3,4-ij:5,6,7-i'j' diquinolizin-18-ium inner salt, hydrogen 9-2,4-disulphonatophenyl-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-xantheno 2,3,4-ij:5,6,7-i'j' diquinolizin-18-ium | |
CHEBI:52274 | |
[O-]S(=O)(=O)C1=CC(=CC=C1C1=C2C=C3CCCN4CCCC(C2=[O+]C2=C5CCCN6CCCC(C=C12)=C56)=C34)S(Cl)(=O)=O |
Specifications
60311-02-6 | |
Glass bottle | |
MFCD00012407 | |
sulforhodamine 101, sulforhodamine 101; sulforhodamine 640, unii-fx0es3271v, texas red, sulforhodamine 101 acid chloride, 2-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-pyrido 3,2,1-ij quinolizino 1',9':6,7,8 chromeno 2,3-f quinolin-4-ium-9-yl-5-sulfobenzenesulfonate, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9$l^ 5 ,23-diazaheptacyclo 17.7.1.1^ 5,9 .0^ 2,17 .0^ 4,15 .0^ 23,27 .0^ 13,28 octacosa-1 27 ,2 17 ,4,9 28 ,13,15,18-heptaen-9-ylium, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9??,23-diazaheptacyclo 17.7.1.1?,?.0 2 , 1 ?.0?, 1 ?.0 2 3 , 2 ?.0 1 3 , 2 ? octacosa-1 27 ,2 17 ,4,9 28 ,13,15,18-heptaen-9-ylium, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9lambda,23-diazaheptacyclo 17.7.1.1,.0 2 , 1 .0, 1 .0 2 3 , 2 .0 1 3 , 2 octacosa-1,4,9 28 ,13,15,17,19 27-heptaen-9-ylium, 9-2,4-disulfophenyl-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-xantheno 2,3,4-ij:5,6,7-i'j' diquinolizin-18-ium inner salt, hydrogen 9-2,4-disulphonatophenyl-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-xantheno 2,3,4-ij:5,6,7-i'j' diquinolizin-18-ium | |
[O-]S(=O)(=O)C1=CC(=CC=C1C1=C2C=C3CCCN4CCCC(C2=[O+]C2=C5CCCN6CCCC(C=C12)=C56)=C34)S(Cl)(=O)=O | |
625.15 | |
CHEBI:52274 | |
99% | |
Sulforhodamine 101 (free acid), (free acid) |
98.5% min. (HPLC) | |
C31H29ClN2O6S2 | |
100 mg | |
MPLHNVLQVRSVEE-UHFFFAOYSA-N | |
16-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3λâ´-oxa-9,23-diazaheptacyclo[17.7.1.1âµ,â¹.0²,¹â·.0â´,¹âµ.0²³,²â·.0¹³,²â¸]octacosa-1,3,5(28),13,15,17,19(27)-heptaen-3-ylium | |
122180 | |
606.71 | |
Pure |
Safety and Handling
EINECSNumber : 262-159-4
RUO – Research Use Only