Sulfonic Acid Polystyrene Resin cross-linked with 15% DVB (70-90mesh) (2.6-3.0mmol/g), TCI America™
Manufacturer:  TCI America S054425G
 View more versions of this product
Catalog No. S054425G
Specifications
Specifications
Sulfonic Acid Polystyrene Resin cross-linked with 15% DVB (70-90mesh) (2.6-3.0mmol/g)
25g
3261
Grain at 20°C
Undesignated
MFCD02683442
Benzenesulfonic Acid Resin
Chemical Identifiers
MFCD02683442
Benzenesulfonic Acid Resin
Safety and Handling
Safety and Handling
TSCA : No
Documents