Learn More
Sulfobromophthalein sodium hydrate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 173400050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sulfobromophthalein sodium hydrate | |
White | |
Glass bottle | |
MFCD16556312 | |
disodium 2-hydroxy-5-1z-4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene 2,3,4,5-tetrabromo-6-carboxyphenyl methyl benzenesulfonate tetrahydrate | |
GHAFORRTMVIXHS-UHFFFAOYSA-L | |
disodium;2-hydroxy-5-[(E)-(4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene)-(2,3,4,5-tetrabromo-6-carboxyphenyl)methyl]benzenesulfonate | |
16211419 | |
≥95% |
123359-42-2 | |
Authentic | |
C20H8Br4Na2O10S2 | |
5 g | |
Solubility in water: soluble | |
[Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C1C(Br)=C(Br)C(Br)=C2Br)C1=CC=C(O)C(=C1)S([O-])(=O)=O | |
837.99 | |
837.76 | |
Crystalline Powder |
Chemical Identifiers
123359-42-2 | |
837.99 | |
GHAFORRTMVIXHS-UHFFFAOYSA-L | |
16211419 | |
[Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C1C(Br)=C(Br)C(Br)=C2Br)C1=CC=C(O)C(=C1)S([O-])(=O)=O |
C20H8Br4Na2O10S2 | |
MFCD16556312 | |
disodium 2-hydroxy-5-1z-4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene 2,3,4,5-tetrabromo-6-carboxyphenyl methyl benzenesulfonate tetrahydrate | |
disodium;2-hydroxy-5-[(E)-(4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene)-(2,3,4,5-tetrabromo-6-carboxyphenyl)methyl]benzenesulfonate |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
In case of inadequate ventilation wear respiratory protecti
GHS Signal Word: Danger
RUO – Research Use Only