Please login to your online account to display your discounted pricing

Sulfo Orange Indicator, 0.04% (w/v) Aqueous Solution, Ricca Chemical

$65.61 - $65.61

Chemical Identifiers

Chemical Name or Material Sulfo Orange Indicator
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 547-57-9
CAS Min % 0.04
CAS Max % 0.04
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8113-16

 
ricca chemical company
RI811316
Glass amber 500mL Each for $65.61

8113-4

 
ricca chemical company
RI81134
Glass amber 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

81135

 
ricca chemical company
RI81135
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear dark reddish orange
Quantity 500mL
Concentration or Composition Notes 0.04% (w/v)
Solvent or Matrix Aqueous Solution
Chemical Name or Material Sulfo Orange Indicator
CAS 7732-18-5
Packaging Glass amber
CAS Max % 100.0
CAS 547-57-9
CAS Max % 0.04
CAS Min % 98.0
CAS Min % 0.04