Please login to your online account to display your discounted pricing

Alfa Aesar™ Sulfated Hirudin (55-65), Certified Mass Spec Grade


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67753LPL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67753LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Sequence DFEEIPEE-Y(SO3H)-LQ
Product Type Sulfated Hirudin (55-65), Certified Mass Spec Grade
Molecular Weight 1491.50
Format Lyophilized powder
Regulatory Status RUO
Purification Method HPLC
Quantity 1 vial, 500 pmol
Storage Requirements -30° to -10°C