Please login to your online account to display your discounted pricing

Sulfate Standard, Certified, 1000ppm ±10ppm (1mL = 1mg SO4), LabChem™


Quantity 1L
Packaging Poly Bottle

Manufacturer: lab chem  LC255102

 View more versions of this product

Catalog No. LC255102

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 142.04
Purity Grade Certified Grade
Solubility Soluble in water
Concentration 1000ppm ±10ppm (1mL = 1mg SO4)
CAS 7757-82-6
CAS Max % 0.15
Packaging Poly Bottle
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 99.85
Name Note 1000ppm (1mL = 1mg SO4)
Identification Passes Test
Viscosity 1.35cSt
Color Clear, colorless
Physical Form Liquid
Quantity 1L
Linear Formula Na2SO4
Chemical Name or Material Sulfate Standard
CAS 7732-18-5
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Sulfate Standard
CAS 7732-18-5
CAS Max % 99.85
CAS 7757-82-6
CAS Max % 0.15
Name Note 1000ppm (1mL = 1mg SO4)
Purity Grade Certified Grade
Linear Formula Na2SO4
Formula Weight 142.04
Accessories

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature