Please login to your online account to display your discounted pricing

Sulfate Standard, 1mL = 0.1mg SO42-, Ricca Chemical

$63.94 - $102.88

Chemical Identifiers

Chemical Name or Material Sulfate Standard
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 7757-82-6
CAS Min % 0.01
CAS Max % 0.02
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8110-16

  SDS
ricca chemical company
RI811016
Poly natural 500mL Each for $63.94

811032

  SDS
ricca chemical company
RI811032
Poly natural 1L Each for $102.88

R8110000120

 
ricca chemical company
RIR8110000120A
Poly natural 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 500mL
Equivalence 1 mL = 0.1 mg SO
CAS 7732-18-5
CAS 7757-82-6
CAS Min % 0.01
CAS Min % 98.0
CAS Max % 100.0
CAS Max % 0.02
Packaging Poly natural
Chemical Name or Material Sulfate Standard