Please login to your online account to display your discounted pricing

Sulfanilic Acid TS, 0.8% (w/v) in Glacial Acetic Acid, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sulfanilic Acid
Name Note Test Solution
CAS 7732-18-5
CAS Min % 72.0
CAS Max % 74.0
CAS 64-19-7
CAS Min % 25.85
CAS Max % 26.91
CAS 121-57-3
CAS Min % 0.57
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8104-16

 
ricca chemical company
RI810416
Glass amber 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

8104-4

 
ricca chemical company
RI81044
Glass amber 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 500mL
Chemical Name or Material Sulfanilic Acid
CAS Max % 0.6
Concentration or Composition Notes 0.8% (w/v)
Solvent or Matrix Glacial Acetic Acid
Name Note Test Solution
CAS 7732-18-5
CAS Min % 72.0
CAS Max % 74.0
Packaging Glass amber
CAS Min % 25.85
CAS Max % 26.91
CAS 121-57-3
CAS Min % 0.57
CAS 64-19-7