Please login to your online account to display your discounted pricing

Sulfanilic acid, ACS reagent, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  419845000

 View more versions of this product

Catalog No. AC419845000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Crystalline Powder
Formula Weight 173.19
Quantity 500g
Color Off-white
Beilstein 14, 695
Merck Index 15, 9058
Assay 98.0 to 102.0%
TSCA TSCA
EINECS Number 204-482-5
RTECS Number WP3895500
Residue after Ignition 0.01% max.
Packaging Plastic bottle
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Synonym 4-Aminobenzenesulfonic acid
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.
Causes skin irritation.
Health Hazard 1 Warning
Insoluble Matter 0.02% max. (in Na2CO3-solution)
Chlorine (Cl) 20ppm max.
Nitrite 0.5ppm max.
Infrared Spectrum Authentic
Sulfate 100ppm max.
Linear Formula (H2N)C6H4SO3H
CAS 121-57-3
Molecular Formula C6H7NO3S
MDL Number MFCD00007886
Chemical Name or Material Sulfanilic acid

Specifications

Chemical Name or Material Sulfanilic acid
CAS 121-57-3
Purity Grade ACS Reagent Grade
Assay 98.0 to 102.0%
Molecular Formula C6H7NO3S
Linear Formula (H2N)C6H4SO3H
Formula Weight 173.19
MDL Number MFCD00007886
Synonym 4-Aminobenzenesulfonic acid
Accessories

Warning

GHS P Statement:
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.
Causes skin irritation.

SDS & Safety Documents