Please login to your online account to display your discounted pricing

Sulfanilic acid, ACS reagent, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  419841000

 View more versions of this product

Catalog No. AC419841000

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Crystalline Powder
Formula Weight 173.19
Quantity 100g
Color Off-white
Beilstein 14, 695
Merck Index 15, 9058
EINECS Number 204-482-5
TSCA TSCA
Assay 98.0 to 102.0%
RTECS Number WP3895500
Infrared Spectrum Authentic
Insoluble Matter 0.02% max. (in Na2CO3-solution)
Synonym 4-Aminobenzenesulfonic acid
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.
Causes skin irritation.
Chemical Name or Material Sulfanilic acid
Chlorine (Cl) 20ppm max.
Molecular Formula C6H7NO3S
Linear Formula (H2N)C6H4SO3H
MDL Number MFCD00007886
CAS 121-57-3
Health Hazard 1 Warning
Nitrite 0.5ppm max.
Residue after Ignition 0.01% max.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Sulfate 100ppm max.

Specifications

Chemical Name or Material Sulfanilic acid
CAS 121-57-3
Purity Grade ACS Reagent Grade
Assay 98.0 to 102.0%
Molecular Formula C6H7NO3S
Linear Formula (H2N)C6H4SO3H
Formula Weight 173.19
MDL Number MFCD00007886
Synonym 4-Aminobenzenesulfonic acid
Accessories

Warning

GHS P Statement:
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.
Causes skin irritation.

SDS & Safety Documents