Please login to your online account to display your discounted pricing

Sulfanilic acid, ACS reagent, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC419840025

 View more versions of this product

Catalog No. AC419840025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 173.19
Physical Form Crystalline Powder
Purity Grade ACS Reagent Grade
Quantity 2.5kg
Color Off-white
Assay 98.0 to 102.0%
RTECS Number WP3895500
EINECS Number 204-482-5
TSCA TSCA
Beilstein 14, 695
Merck Index 15, 9058
Synonym 4-Aminobenzenesulfonic acid
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Insoluble Matter 0.02% max. (in Na2CO3-solution)
Packaging Plastic bottle
Residue after Ignition 0.01% max.
Nitrite 0.5ppm max.
Sulfate 100ppm max.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.
Causes skin irritation.
Chlorine (Cl) 20ppm max.
Health Hazard 1 Warning
Chemical Name or Material Sulfanilic acid
Linear Formula (H2N)C6H4SO3H
Molecular Formula C6H7NO3S
CAS 121-57-3
MDL Number MFCD00007886

Specifications

Chemical Name or Material Sulfanilic acid
CAS 121-57-3
Purity Grade ACS Reagent Grade
Assay 98.0 to 102.0%
Molecular Formula C6H7NO3S
Linear Formula (H2N)C6H4SO3H
Formula Weight 173.19
MDL Number MFCD00007886
Synonym 4-Aminobenzenesulfonic acid
Accessories

Warning

GHS P Statement:
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.
Causes skin irritation.

SDS & Safety Documents