Learn More
Sulfanilic acid, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Reagent for chromatographic detection of histidine
Supplier: Thermo Scientific Chemicals 04154030
Description
Sulfanilic acid is used as a reagent for chromatographic detection of histidine. It is also used as a chemical intermediate in organic synthesis. It is utilized in quantitative analysis of nitrate and nitrite ions. It plays an essential role in the manufacturing of azo dyes and in synthesis of sulfa drugs. It is used as a dopant for the chemical synthesis of polyaniline. It is used to prepare 2,4-diamino-6-p-sulphoanilinopyrimidine by reacting with 6-chloro-pyrimidine-2,4-diamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sulfanilic acid | |
∼365°C | |
MFCD00007886 | |
908765 | |
sulfanilic acid, sulphanilic acid, p-aminobenzenesulfonic acid, sulfanilsaeure, aniline-4-sulfonic acid, benzenesulfonic acid, 4-amino, aniline-p-sulfonic acid, aniline-p-sulphonic acid, p-aminophenylsulfonic acid, sulfanilicacid | |
HVBSAKJJOYLTQU-UHFFFAOYSA-N | |
4-aminobenzenesulfonic acid | |
8479 | |
173.19 | |
ACS |
121-57-3 | |
C6H7NO3S | |
250 g | |
14,8926 | |
Slightly soluble in water. Insoluble in ethanol,ether,caustic soda and sodium carbonate. | |
C1=CC(=CC=C1N)S(=O)(=O)O | |
173.186 | |
CHEBI:27500 | |
98.0 to 102.0% | |
Powder |
Chemical Identifiers
121-57-3 | |
173.186 | |
HVBSAKJJOYLTQU-UHFFFAOYSA-N | |
8479 | |
4-aminobenzenesulfonic acid |
C6H7NO3S | |
MFCD00007886 | |
sulfanilic acid, sulphanilic acid, p-aminobenzenesulfonic acid, sulfanilsaeure, aniline-4-sulfonic acid, benzenesulfonic acid, 4-amino, aniline-p-sulfonic acid, aniline-p-sulphonic acid, p-aminophenylsulfonic acid, sulfanilicacid | |
CHEBI:27500 | |
C1=CC(=CC=C1N)S(=O)(=O)O |
Safety and Handling
GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
H315-H317-H319
EINECSNumber : 204-482-5
RTECSNumber : WP3895500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only