Please login to your online account to display your discounted pricing

Sulfanilic acid, 99%, ACROS Organics™

$80.70 - $80.70

Chemical Identifiers

Chemical Name or Material Sulfanilic acid
Assay ≥98.5% (Acid)
Assay Percent Range 99%
Molecular Formula C6H7NO3S
Linear Formula (H2N)C6H4SO3H
Formula Weight 173.19
MDL Number MFCD00007886
Synonym 4-Aminobenzenesulfonic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC150710010

  SDS
fisher scientific acros
150710010
Plastic bottle 1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC150711000

  SDS
fisher scientific acros
150711000
Plastic bottle 100g Each for $80.70

AC150710025

  SDS
thermo fisher scientific
AC150710025
Plastic bottle 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC150710100

  SDS
thermo fisher scientific
AC150710100
Plastic drum 10kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5% (Acid)
Physical Form Liquid
Formula Weight 173.19
Quantity 1kg
Color Clear
Assay Percent Range 99%
Synonym 4-Aminobenzenesulfonic acid
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.
Causes skin irritation.
Molecular Formula C6H7NO3S
Linear Formula (H2N)C6H4SO3H
MDL Number MFCD00007886
Infrared Spectrum Authentic
Packaging Plastic bottle
Water 0.2% max. (K.F.)
Chemical Name or Material Sulfanilic acid
CAS 121-57-3
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.

Safety and Handling

Warning

GHS P Statement:
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.
Causes skin irritation.