Please login to your online account to display your discounted pricing

Sulfanilamide, Reagent Grade, >98.0%, LabChem™

$41.71 - $41.71

Chemical Identifiers

Chemical Name or Material Sulfanilamide
CAS 63-74-1
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C6H8N2O2S
Formula Weight 172.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC254409

 
lab chem
LC254409
Amber Glass 100g Each for $41.71
Description & Specifications

Specifications

Formula Weight 172.2
CAS 63-74-1
Identification Passes Test
Purity Grade Reagent Grade
Packaging Amber Glass
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
EINECS Number 200-563-4
Density 1.08g/cm3
Physical Form Powder
Assay >98.0%
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Linear Formula C6H8N2O2S
Recommended Storage Room Temperature
Quantity 100g
Health Hazard 1 Warning
Specific Gravity 1.08g/cm3
Color White
Chemical Name or Material Sulfanilamide
Solubility Information Soluble in water
CAS Max % 100
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Room Temperature