Please login to your online account to display your discounted pricing

Sulfanilamide/N-(Naphthyl) Reagent, For Nitrogen (Nitrate/Nitrite), LabChem™

$102.11 - $175.57

Chemical Identifiers

Chemical Name or Material Sulfanilamide/N-(Naphthyl) Reagent
CAS 7732-18-5
CAS Max % 85.39
CAS 7664-38-2
CAS Max % 13.51
CAS 63-74-1
CAS Max % 1
CAS 1465-25-4
CAS Max % 0.1
Name Note For Nitrogen (Nitrate/Nitrite)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC254601

 
lab chem
LC254601
Amber Poly 500mL Each for $102.11

LC254602

 
lab chem
LC254602
Amber Poly 1L Each for $175.57
Description & Specifications

Specifications

Formula Weight 259.18
Viscosity 1.35cSt
CAS Max % 1
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds; Phosphorus oxides
Color Clear, slight pink to tan
Quantity 500mL
CAS Max % 0.1
Name Note For Nitrogen (Nitrate/Nitrite)
Physical Form Liquid
Linear Formula C10H7NHCH2CH2NH2·2HCl
Density 1.07g/mL
CAS 7732-18-5
UN Number UN1805
Health Hazard 1 Danger
CAS Max % 85.39
CAS 7664-38-2
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Amber Poly
CAS Max % 13.51
Solubility Soluble in water
CAS 63-74-1
Specific Gravity 1.07g/mL
Storage Information Refrigerate
Chemical Name or Material Sulfanilamide/N-(Naphthyl) Reagent
CAS 1465-25-4
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Identification Passes Test
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.