Please login to your online account to display your discounted pricing

Sulfanilamide, BAKER™, J.T.Baker™

A grade of chemicals for laboratory use

$154.76 - $248.84

Chemical Identifiers

Chemical Name or Material Sulfanilamide
CAS 63-74-1
Molecular Formula 4-NH2C6H4SO2NH2
Formula Weight 172.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004400

 
avantor performance
V15305
Wide Mouth Amber Glass Bottle 100g Each for $154.76

02004401

 
avantor performance
V15307
Wide Mouth Amber Glass Bottle 500g Each for $248.84
Description & Specifications

Specifications

Formula Weight 172.21
Chemical Name or Material Sulfanilamide
Molecular Formula 4-NH2C6H4SO2NH2
CAS 63-74-1
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.08kg
Quantity 100g