missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfamide, 98+%, ACROS Organics™

Manufacturer:  Acros Organics 199020250

 View more versions of this product

Catalog No. AC199020250



Specifications

Specifications

Sulfamide
98.0% min.
0.5% (50°C, 2 hrs) (vacuum) max.
1.61
87.0°C to 91.0°C
5.0 to 7.0 (10% soln. aq. soln.)
98+%
H2NSO2NH2
imidosulfamic acid
NVBFHJWHLNUMCV-UHFFFAOYSA-N
sulfamide
82267
96.11
98+%
1.6100g/mL
Authentic
Glass bottle
White
25g
7803-58-9
H4N2O2S
15,9054
Solubility in water: soluble. Other solubilities: soluble in acetone,hot alcohol
NS(=O)(=O)N
96.11
CHEBI:29368
Crystalline Powder, Free From Visible Impurities

Chemical Identifiers

7803-58-9
96.11
imidosulfamic acid
CHEBI:29368
NS(=O)(=O)N
H4N2O2S
NVBFHJWHLNUMCV-UHFFFAOYSA-N
82267
sulfamide
Safety and Handling

Safety and Handling

Warning

SDS Documents