Please login to your online account to display your discounted pricing

Sulfamide, 97%, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  SB01649EA

 View more versions of this product

Catalog No. SB01649EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 96.11
Quantity 10g
Assay 97%
CAS Min % 97.0
Linear Formula H2NSO2NH2
CAS 7803-58-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Sulfamide
Assay Percent Range 97%
Molecular Formula H4N2O2S
MDL Number MFCD00011606

Specifications

Chemical Name or Material Sulfamide
CAS 7803-58-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula H4N2O2S
Linear Formula H2NSO2NH2
Formula Weight 96.11
MDL Number MFCD00011606
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.