Please login to your online account to display your discounted pricing

Sulfamic Acid , Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Sulfamic Acid
Assay Percent Range 99%
Molecular Formula NH2SO3H
Formula Weight 97.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK1931212

  SDS
avantor performance
193105
Glass 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK1931500

 
avantor performance
193159
Glass 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 97.09
CAS 5329-14-6
Molecular Formula NH2SO3H
Chemical Name or Material Sulfamic Acid
Assay Percent Range 99%
Packaging Glass
Quantity 2.5kg