Please login to your online account to display your discounted pricing

Sulfamic Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$141.58 - $332.64

Chemical Identifiers

Chemical Name or Material Sulfamic Acid
CAS 5329-14-6
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula NH2SO3H
Formula Weight 97.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004350

 
avantor performance
V14507
Poly Bottle 500g Each for $332.64

02004347

 
avantor performance
V14505
Poly Bottle 100g Each for $141.58
Description & Specifications

Specifications

Formula Weight 97.09
Purity Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Sulfamic Acid
CAS 5329-14-6
Quantity 500g
Molecular Formula NH2SO3H
Specific Gravity 1L = 2.10kg
Packaging Poly Bottle