Please login to your online account to display your discounted pricing

Sulfamic Acid, 99.3 to 100.3%, EMD Millipore

$347.17 - $347.17

Chemical Identifiers

Chemical Name or Material Sulfamic Acid
CAS 5329-14-6
Name Note For Synthesis
Molecular Formula H3NO3S
Linear Formula HCH2NSOCH3H
Formula Weight 97.8
Synonym Amidosulfonic acid
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

MSX11403

 
emd millipore corporation
SX11403
Each for $347.17
Description & Specifications

Specifications

Physical Form Crystals
Formula Weight 97.8
Color White
Chemical Name or Material Sulfamic Acid
Vapor Pressure 0.025 hPa (100°C)
Name Note For Synthesis
Bulk Density 600kg/m3
Assay Percent Range 99.3 to 100.3%
Molecular Formula H3NO3S
Linear Formula HCH2NSOCH3H
CAS 5329-14-6
Synonym Amidosulfonic acid
Recommended Storage Store at +15°C to +25°C
Packaging Plastic Bottle
Safety and Handling

Store at +15°C to +25°C