missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfamethazine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1927622

 View more versions of this product

Catalog No. AAA1927622



Description

Description

Sulfamethazine blocks the synthesis of dihydrofolic acid by inhibiting dihydropteroate synthase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfamethazine is an antibacterial. Sulfamethazine blocks the synthesis of dihydrofolic acid by inhibiting dihydropteroate synthase.Sulfamethazine is an antibiotic used to treat bronchitis, prostatitis and urinary tract infections. It is used in disposition and depletion kinetic studies. It is used to develop detection techniques for quantification in fluids such as cows’ milk, honey and swine urine. Sulfamethazine is an antimicrobial sulfur drug that blocks the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase. Sulfamethazine is a competitive inhibitor of bacterial para-aminobenzoic acid.

Solubility
Soluble in DMSO (56 mg/mL at 25°C), water (<1mg/mL at 25°C), and ethanol (<1mg/mL at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

Sulfamethazine
198°C to 201°C
MFCD00006066
sulfamethazine, sulfadimidine, sulfadimethyldiazine, sulfamezathine, sulfadimerazine, sulphamethazine, sulphamezathine, sulfadimezine, sulphadimidine, sulfadimesin
ASWVTGNCAZCNNR-UHFFFAOYSA-N
4-amino-N-(4,6-dimethylpyrimidin-2-yl)benzenesulfonamide
5327
278.34
57-68-1
C12H14N4O2S
100 g
Soluble in DMSO (56mg/mL at 25°C),water (<1mg/mL at 25°C),and ethanol (<1mg/mL at 25°C).
CC1=CC(=NC(=N1)NS(=O)(=O)C2=CC=C(C=C2)N)C
278.33
CHEBI:102265
99%

Chemical Identifiers

57-68-1
278.33
ASWVTGNCAZCNNR-UHFFFAOYSA-N
5327
4-amino-N-(4,6-dimethylpyrimidin-2-yl)benzenesulfonamide
C12H14N4O2S
MFCD00006066
sulfamethazine, sulfadimidine, sulfadimethyldiazine, sulfamezathine, sulfadimerazine, sulphamethazine, sulphamezathine, sulfadimezine, sulphadimidine, sulfadimesin
CHEBI:102265
CC1=CC(=NC(=N1)NS(=O)(=O)C2=CC=C(C=C2)N)C
Safety and Handling

Safety and Handling

EINECSNumber : 200-346-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only