Please login to your online account to display your discounted pricing

Alfa Aesar™ Sulfamerazine, 98+%

$29.24 - $29.24

Chemical Identifiers

Chemical Name or Material Sulfamerazine
CAS 127-79-7
Molecular Formula C11H12N4O2S
Formula Weight 264.3
MDL Number MFCD00023212
Beilstein 249133
Merck Index 14,8913
Synonym N(1)-(4-Methyl-2-pyrimidinyl)sulfanilamide; 2-Sulfanilamido-4-methylpyrimidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0419414

 
alfa aesar™
L0419414
25g Each for $29.24
Description & Specifications

Specifications

Formula Weight 264.3
Chemical Name or Material Sulfamerazine
Assay ≥98%
MDL Number MFCD00023212
CAS 127-79-7
Molecular Formula C11H12N4O2S
Beilstein 249133
Synonym N(1)-(4-Methyl-2-pyrimidinyl)sulfanilamide; 2-Sulfanilamido-4-methylpyrimidine
RTECS Number WP0750000
Quantity 25g
TSCA TSCA
Merck Index 14,8913
Melting Point 234° to 238°C