missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfalene 98.0+%, TCI America™

Manufacturer:  TCI America S091725G

 View more versions of this product

Catalog No. S091725G



Specifications

Specifications

Sulfalene
178°C
152-47-6
MFCD00437754
KXRZBTAEDBELFD-UHFFFAOYSA-N
4-amino-N-(3-methoxypyrazin-2-yl)benzenesulfonamide
9047
280.30
≥98.0% (HPLC,T)
White
25g
C11H12N4O3S
4-Amino-N-(3-methoxypyrazin-2-yl)benzenesulfonamide, N-(3-Methoxypyrazin-2-yl)sulfanilamide
COC1=NC=CN=C1NS(=O)(=O)C2=CC=C(C=C2)N
280.302
CHEBI:32162
Crystal-Powder at 20°C

Chemical Identifiers

152-47-6
280.302
KXRZBTAEDBELFD-UHFFFAOYSA-N
9047
4-amino-N-(3-methoxypyrazin-2-yl)benzenesulfonamide
C11H12N4O3S
MFCD00437754
4-Amino-N-(3-methoxypyrazin-2-yl)benzenesulfonamide, N-(3-Methoxypyrazin-2-yl)sulfanilamide
CHEBI:32162
COC1=NC=CN=C1NS(=O)(=O)C2=CC=C(C=C2)N
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0767

RTECSNumber : WP0175000

TSCA : No

Documents