missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sulfalene 98.0+%, TCI America™
$189.13 - $474.48
Chemical Identifiers
CAS | 152-47-6 |
---|---|
Molecular Formula | C11H12N4O3S |
Molecular Weight (g/mol) | 280.302 |
MDL Number | MFCD00437754 |
InChI Key | KXRZBTAEDBELFD-UHFFFAOYSA-N |
Synonym | 4-Amino-N-(3-methoxypyrazin-2-yl)benzenesulfonamide, N-(3-Methoxypyrazin-2-yl)sulfanilamide |
PubChem CID | 9047 |
ChEBI | CHEBI:32162 |
IUPAC Name | 4-amino-N-(3-methoxypyrazin-2-yl)benzenesulfonamide |
SMILES | COC1=NC=CN=C1NS(=O)(=O)C2=CC=C(C=C2)N |
Chemical Identifiers
152-47-6 | |
280.302 | |
KXRZBTAEDBELFD-UHFFFAOYSA-N | |
9047 | |
4-amino-N-(3-methoxypyrazin-2-yl)benzenesulfonamide |
C11H12N4O3S | |
MFCD00437754 | |
4-Amino-N-(3-methoxypyrazin-2-yl)benzenesulfonamide, N-(3-Methoxypyrazin-2-yl)sulfanilamide | |
CHEBI:32162 | |
COC1=NC=CN=C1NS(=O)(=O)C2=CC=C(C=C2)N |
Specifications
152-47-6 | |
White | |
MFCD00437754 | |
4-Amino-N-(3-methoxypyrazin-2-yl)benzenesulfonamide, N-(3-Methoxypyrazin-2-yl)sulfanilamide | |
COC1=NC=CN=C1NS(=O)(=O)C2=CC=C(C=C2)N | |
280.302 | |
CHEBI:32162 | |
≥98.0% (HPLC,T) | |
Sulfalene |
178°C | |
C11H12N4O3S | |
5 g | |
KXRZBTAEDBELFD-UHFFFAOYSA-N | |
4-amino-N-(3-methoxypyrazin-2-yl)benzenesulfonamide | |
9047 | |
280.30 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-0767
RTECSNumber : WP0175000
TSCA : No