missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfadiazine, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1237018

 View more versions of this product

Catalog No. AAA1237018



Description

Description

Solubility
Soluble in dimethyl sulfoxide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Sulfadiazine
68-35-9
C10H10N4O2S
14,8903
SEEPANYCNGTZFQ-UHFFFAOYSA-N
4-amino-N-pyrimidin-2-ylbenzenesulfonamide
5215
250.28
50g
99%
MFCD00006065
sulfadiazine, sulphadiazine, sulfapyrimidine, sulfadiazin, sulfazine, sulfadiazene, liquadiazine, sulfapyrimidin, adiazin, adiazine
C1=CN=C(N=C1)NS(=O)(=O)C2=CC=C(C=C2)N
250.276
CHEBI:9328
99%

Chemical Identifiers

68-35-9
250.276
SEEPANYCNGTZFQ-UHFFFAOYSA-N
5215
4-amino-N-pyrimidin-2-ylbenzenesulfonamide
C10H10N4O2S
MFCD00006065
sulfadiazine, sulphadiazine, sulfapyrimidine, sulfadiazin, sulfazine, sulfadiazene, liquadiazine, sulfapyrimidin, adiazin, adiazine
CHEBI:9328
C1=CN=C(N=C1)NS(=O)(=O)C2=CC=C(C=C2)N
Safety and Handling

Safety and Handling

GHS H Statement
H334-H302-H335-H315-H319-H317

GHS P Statement
P284a-P261-P280-P305+P351+P338-P304+P340-P342+P311-P362-P301+P312-P405-P501a

Danger

RTECSNumber : WP1925000

TSCA : TSCA

SDS Documents