Please login to your online account to display your discounted pricing

Alfa Aesar™ Sulfacetamide sodium salt hydrate, 99%

$37.36 - $337.51

Chemical Identifiers

Chemical Name or Material Sulfacetamide sodium salt hydrate
CAS 6209-17-2
Molecular Formula C8H9N2NaO3S·xH2O
Formula Weight 236.23 (Anhydrous)
MDL Number MFCD00007885
Synonym N-(4-Aminobenzenesulfonyl)acetamide sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2242614

  SDS
alfa aesar™
B2242614
25g Each for $37.36

AAB2242622

  SDS
alfa aesar™
B2242622
100g Each for $89.80

AAB2242636

  SDS
alfa aesar™
B2242636
500g Each for $337.51
Description & Specifications

Specifications

Formula Weight 236.23 (Anhydrous)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Sulfacetamide sodium salt hydrate
RTECS Number AC8480000
Molecular Formula C8H9N2NaO3S·xH2O
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00007885
Quantity 25g
Assay 99%
CAS 6209-17-2
Health Hazard 1 Warning
Synonym N-(4-Aminobenzenesulfonyl)acetamide sodium salt

Applications
Sulfacetamide sodium salt hydrate is an anti-infective agent that is used topically to treat skin infections and orally for urinary tract infections. It adapts to cure trachoma, conjunctivitis, keratitis.

Solubility
Freely soluble in water. Insoluble in chloroform, ether, and benzene.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.