Please login to your online account to display your discounted pricing

Alfa Aesar™ Sulfacetamide, 98%

$112.86 - $112.86

Chemical Identifiers

Chemical Name or Material Sulfacetamide
CAS 144-80-9
Molecular Formula C8H10N2O3S
Formula Weight 214.24
MDL Number MFCD00066501
Beilstein 981718
Merck Index 14,8899
Synonym N-(4-Aminobenzenesulfonyl)acetamide; N-Sulfanilylacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1983630

  SDS
alfa aesar™
A1983630
250g Each for $112.86

AAA1983618

  SDS
alfa aesar™
A1983618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sulfacetamide
Quantity 250g
CAS 144-80-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H341
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number AC8450000
TSCA TSCA
Assay 98%
Molecular Formula C8H10N2O3S
Formula Weight 214.24
MDL Number MFCD00066501
Beilstein 981718
Merck Index 14,8899
Synonym N-(4-Aminobenzenesulfonyl)acetamide; N-Sulfanilylacetamide
Melting Point 182° to 184°C
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341
Suspected of causing genetic defects.