missing translation for 'onlineSavingsMsg'
Learn More

Sulbactam 98.0+%, TCI America™

Supplier:  TCI America S086825G

 View more versions of this product

Catalog No. S086825G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sulbactam
156°C
C8H11NO5S
25 g
FKENQMMABCRJMK-RITPCOANSA-N
(2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
130313
233.24
Crystalline Powder
68373-14-8
White-Yellow
MFCD00867005
sulbactam, sulbactam acid, penicillanic acid sulfone, sulbactamum, betamaze, 2s,5r-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid 4,4-dioxide, penicillanic acid 1,1-dioxide, unii-s4tf6i2330, penicillanic acid dioxide, chembl403
CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O
233.24
CHEBI:9321
≥98.0% (HPLC,T)

Chemical Identifiers

68373-14-8
233.24
FKENQMMABCRJMK-RITPCOANSA-N
130313
(2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
C8H11NO5S
MFCD00867005
sulbactam, sulbactam acid, penicillanic acid sulfone, sulbactamum, betamaze, 2s,5r-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid 4,4-dioxide, penicillanic acid 1,1-dioxide, unii-s4tf6i2330, penicillanic acid dioxide, chembl403
CHEBI:9321
CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions