Please login to your online account to display your discounted pricing

Alfa Aesar™ Sudan II

$121.59 - $121.59

Chemical Identifiers

Chemical Name or Material Sudan II
CAS 3118-97-6
Molecular Formula C18H16N2O
Formula Weight 276.34
MDL Number MFCD00003896
Synonym C.I. 12140; Solvent Orange 7
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1761322

  SDS
alfa aesar™
A1761322
100g Each for $121.59
Description & Specifications

Specifications

Formula Weight 276.34
CAS 3118-97-6
Melting Point 156° to 160°C
TSCA TSCA
Chemical Name or Material Sudan II
Molecular Formula C18H16N2O
Health Hazard 2 GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
MDL Number MFCD00003896
Health Hazard 3 GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 100g
Synonym C.I. 12140; Solvent Orange 7
Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.