missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sucrose octabenzoate, 98%

$49.66 - $97.61

Chemical Identifiers

CAS 12738-64-6
Molecular Formula C19H28O13
Molecular Weight (g/mol) 464.42
MDL Number MFCD00047542
InChI Key GYUJITDJMFVEPZ-CKQFFSBTNA-N
Synonym Sucrose benzoate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6608322 View Documents Alfa Aesar™
J6608322
100g Each for $49.66
 
AAJ6608336 View Documents Alfa Aesar™
J6608336
500g Each for $97.61
 
Specifications

Chemical Identifiers

12738-64-6
464.42
GYUJITDJMFVEPZ-CKQFFSBTNA-N
C19H28O13
MFCD00047542
Sucrose benzoate

Specifications

White
100g
C19H28O13
Sucrose benzoate
464.42
Powder
Sucrose octabenzoate
93°C to 100°C
12738-64-6
MFCD00047542
GYUJITDJMFVEPZ-CKQFFSBTNA-N
1175.14
98%
Safety and Handling

Safety and Handling

TSCA : TSCA