missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sucrose NF GenAR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Formula Weight 342.3
Chemical Name or Material Sucrose
Grade NF
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK77231 Avantor Performance
772304
POLYSTORMOR 1kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

57-50-1
342.3
NF
C12H22O11
Sucrose

Specifications

342.3
POLYSTORMOR
1kg
NF
C12H22O11
57-50-1
Sucrose