Please login to your online account to display your discounted pricing

Sucrose, High Purity (Low Endotoxin), Multi-Compendial, N.F., J.T.Baker™

$211.62 - $1,389.60

Chemical Identifiers

Chemical Name or Material Sucrose
CAS 57-50-1
Name Note High Purity (Low Endotoxin)
Purity Grade Multi-Compendial/NF
Molecular Formula C12H22O11
Formula Weight 342.3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004327

 
avantor performance
400504
Poly Bottle 100g Each for $211.62

02004330

 
avantor performance
400505
Poly Bottle 1kg Each for $1,389.60
Description & Specifications

Specifications

Purity Grade Multi-Compendial/NF
Formula Weight 342.3
Chemical Name or Material Sucrose
Molecular Formula C12H22O11
CAS 57-50-1
Specific Gravity 1L = 1.59kg
Quantity 100g
Name Note High Purity (Low Endotoxin)
Packaging Poly Bottle