Please login to your online account to display your discounted pricing

Sucrose, High Purity (Low Endotoxin), Multi-Compendial, N.F., J.T.Baker™


Quantity 1kg
Packaging Poly Bottle

Manufacturer: avantor performance  400505

 View more versions of this product

Catalog No. 02004330

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 342.3
Purity Grade Multi-Compendial/NF
Chemical Name or Material Sucrose
CAS 57-50-1
Molecular Formula C12H22O11
Name Note High Purity (Low Endotoxin)
Packaging Poly Bottle
Specific Gravity 1L = 1.59kg
Quantity 1kg

Specifications

Chemical Name or Material Sucrose
CAS 57-50-1
Name Note High Purity (Low Endotoxin)
Purity Grade Multi-Compendial/NF
Molecular Formula C12H22O11
Formula Weight 342.3