Please login to your online account to display your discounted pricing

Sucrose gel loading dye, 5x, ACROS Organics™


Quantity 5mL

Manufacturer: fisher scientific acros  612000050

 View more versions of this product

Catalog No. AC612000050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 5mL
DNase DNase free
Absorbance 0.30 - 0.45 AU at 252nm
Chemical Name or Material Sucrose gel loading dye
Electrophoresis Pass Test
CAS 57-50-1

Specifications

Chemical Name or Material Sucrose gel loading dye
CAS 57-50-1