missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sucrose gel loading dye, 5x, ACROS Organics™

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.297
InChI Key CZMRCDWAGMRECN-UGDNZRGBSA-N
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC612000010 SDS View Documents Acros Organics
612000010
1mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

57-50-1
342.297
5988
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
C12H22O11
CZMRCDWAGMRECN-UGDNZRGBSA-N
CHEBI:17992
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O

Specifications

0.30 - 0.45 AU at 252nm, 0.47 - 0.62 AU, 0.50 - 0.65 AU
DNase free
C12H22O11
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O
342.297
CHEBI:17992
Sucrose gel loading dye
57-50-1
Pass Test
CZMRCDWAGMRECN-UGDNZRGBSA-N
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
5988
1mL