Please login to your online account to display your discounted pricing

Sucrose (Crystalline/NF), Fisher Chemical

$206.71 - $1,257.80

Chemical Identifiers

Chemical Name or Material Sucrose
CAS 57-50-1
Purity Grade NF
Physical Form Solid
Molecular Formula C12H22O11
Formula Weight 342.3
MDL Number MFCD00006626
Synonym Cane Sugar
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S3-12

  SDS
fisher scientific company
FLS312
Poly Pail 12kg Each for $1,257.80

S3-212

  SDS
fisher scientific company
FLS3212
Poly Bottle 2.5kg Each for $399.33
Case of 4 Each for $1,597.32

S3-500

  SDS
fisher scientific company
FLS3500
Poly Bottle 500g Each for $206.71
Case of 6 Each for $1,240.26
Description & Specifications

Specifications

Melting Point 190°C
Formula Weight 342.3
Quantity 12kg
Physical Form Solid
Purity Grade NF
Molecular Formula C12H22O11
Color White
pH 6.5 to 7.5
Heavy Metals (as Pb) 5ppm max. (NF)
Residual Solvents Meets Requirements (NF)
Chloride 0.0035% max. (NF)
Residue after Ignition 0.05% max. (NF)
MDL Number MFCD00006626
ChemAlert Storage Symbol Gray
Health Hazard 2 CAUTION!
Sulfate 0.006% max. (NF)
Calcium (Ca) Pass Test (NF)
Identification Pass Test (NF)
Invert Sugars 112mg CuO max. (NF)
Packaging Poly Pail
Specific Rotation 65.9° min. (NF)
Chemical Name or Material Sucrose
Synonym Cane Sugar
Health Hazard 3 Emergency Overview
May cause skin, eye, and respiratory tract irritation. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:1
Instability:0
CAS 57-50-1

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:1
Instability:0

CAUTION!