Please login to your online account to display your discounted pricing

Sucrose (Crystalline/NF), Fisher Chemical


Quantity 500g
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLS3500

 View more versions of this product

Catalog No. S3-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 342.3
Melting Point 190°C
Quantity 500g
Physical Form Solid
Purity Grade NF
Molecular Formula C12H22O11
Color White
pH 6.5 to 7.5
Calcium (Ca) Pass Test (NF)
Chloride 0.0035% max. (NF)
Identification Pass Test (NF)
Heavy Metals (as Pb) 5ppm max. (NF)
MDL Number MFCD00006626
ChemAlert Storage Symbol Gray
Synonym Cane Sugar
Invert Sugars 112mg CuO max. (NF)
Packaging Poly Bottle
Health Hazard 2 CAUTION!
Residue after Ignition 0.05% max. (NF)
Residual Solvents Meets Requirements (NF)
Health Hazard 3 Emergency Overview
May cause skin, eye, and respiratory tract irritation. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:1
Instability:0
Specific Rotation 65.9° min. (NF)
Sulfate 0.006% max. (NF)
Chemical Name or Material Sucrose
CAS 57-50-1

Specifications

Chemical Name or Material Sucrose
CAS 57-50-1
Purity Grade NF
Physical Form Solid
Molecular Formula C12H22O11
Formula Weight 342.3
MDL Number MFCD00006626
Synonym Cane Sugar
Accessories
 

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:1
Instability:0

CAUTION!

SDS & Safety Documents