Learn More

Sucrose (Crystalline/NF/EP/BP/JP), Fisher Chemical™
Manufacturer: Fisher Chemical S6500
Specifications
Pass Test (BP/EP/JP) | |
Sucrose | |
0.05% max. (NF) | |
Poly Bottle | |
+66.3 to +67.0° (BP/EP/JP), 65.9° min. (NF) | |
190°C | |
6.5 to 7.5 | |
C12H22O11 | |
amerfand, cane sugar, d-sucrose, rohrzucker, saccharose, saccharum, sucrose, sugar, table sugar, white sugar | |
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O | |
342.297 | |
CHEBI:17992 | |
Solid |
Pass Test (BP/EP/JP) | |
Pass Test (BP/EP/JP/NF) | |
0.1% max. (BP/EP/JP) | |
Meets Requirements (BP/EP/NF) | |
White | |
500g | |
57-50-1 | |
MFCD00006626 | |
CZMRCDWAGMRECN-UGDNZRGBSA-N | |
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
5988 | |
342.3 | |
BP/EP/JP/NF |
Chemical Identifiers
57-50-1 | |
342.297 | |
CZMRCDWAGMRECN-UGDNZRGBSA-N | |
5988 | |
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
C12H22O11 | |
MFCD00006626 | |
amerfand, cane sugar, d-sucrose, rohrzucker, saccharose, saccharum, sucrose, sugar, table sugar, white sugar | |
CHEBI:17992 | |
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O |
Safety and Handling

CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.
NFPA
Health:1
Flammability:1
Instability:0