missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sucrose (Crystalline/NF/EP/BP/JP), Fisher Chemical™

Manufacturer:  Fisher Chemical S6212

 View more versions of this product

Catalog No. S6212



Description

Description

Specifications

Specifications

Pass Test (BP/EP/JP)
Sucrose
0.05% max. (NF)
Poly Bottle
+66.3 to +67.0° (BP/EP/JP), 65.9° min. (NF)
190°C
6.5 to 7.5
C12H22O11
amerfand, cane sugar, d-sucrose, rohrzucker, saccharose, saccharum, sucrose, sugar, table sugar, white sugar
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O
342.297
CHEBI:17992
Solid
Pass Test (BP/EP/JP)
Pass Test (BP/EP/JP/NF)
0.1% max. (BP/EP/JP)
Meets Requirements (BP/EP/NF)
Undesignated
2.5kg
57-50-1
MFCD00006626
CZMRCDWAGMRECN-UGDNZRGBSA-N
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
5988
342.3
BP/EP/JP/NF

Chemical Identifiers

57-50-1
342.297
CZMRCDWAGMRECN-UGDNZRGBSA-N
5988
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
C12H22O11
MFCD00006626
amerfand, cane sugar, d-sucrose, rohrzucker, saccharose, saccharum, sucrose, sugar, table sugar, white sugar
CHEBI:17992
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:1
Instability:0

SDS Documents
Product Certifications