Please login to your online account to display your discounted pricing

Sucrose (Crystalline/NF/EP/BP/JP), Fisher Chemical


Quantity 50kg
Packaging Poly Drum

Manufacturer: fisher scientific company  S650*

 View more versions of this product

Catalog No. S650

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade BP/EP/JP/NF
Formula Weight 342.3
Melting Point 190°C
Quantity 50kg
Physical Form Solid
Color White
Conductivity 35 μS/cm max. (EP)
Molecular Formula C12H22O11
pH 6.5 to 7.5
Residual Solvents Meets Requirements (BP)
Specific Rotation +66.3 to +67.0° (BP)
Sulfite 10ppm max. (BP)
Clarity and Color Pass Test (JP)
Identification Pass Test (BP)
Endotoxin 0.25 EU/mg max. (JP)
Heavy Metals (as Pb) 5ppm max. (NF)
Glucose and Invert Sugars 0.04% max. (BP)
Lead (Pb) 0.5ppm max. (BP)
Loss on Drying 0.1% max. (BP)
Packaging Poly Drum
Appearance of Solution Pass Test (BP)
Calcium (Ca) Pass Test (NF)
Dextrins Pass Test (BP)
MDL Number MFCD00006626
Acidity Alkalinity Pass Test (BP)
CAS 57-50-1
Invert Sugars 0.04% max. (JP)
Health Hazard 2 CAUTION!
Reducing Sugars Pass Test (BP)
Sulfate 0.006% max. (NF)
Residue after Ignition 0.05% max. (NF)
Bacterial Endotoxins Report IU/mg (EP)
Synonym Cane Sugar
Chloride 0.0035% max. (NF)
Chemical Name or Material Sucrose
ChemAlert Storage Symbol Gray
Health Hazard 3 Emergency Overview
May cause skin, eye, and respiratory tract irritation. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:1
Instability:0

Specifications

Chemical Name or Material Sucrose
CAS 57-50-1
Purity Grade BP/EP/JP/NF
Physical Form Solid
Molecular Formula C12H22O11
Formula Weight 342.3
MDL Number MFCD00006626
Synonym Cane Sugar
Accessories

Accessories for Catalog Number: S650

Item Mfr. No. Item Description Price Per Unit Quantity  
Accessories
 

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:1
Instability:0

CAUTION!

SDS & Safety Documents