missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sucrose ACS AR Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 57-50-1
Physical Form Crystalline
Molecular Formula C12H22O11
Formula Weight 342.3
Chemical Name or Material Sucrose
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK8360212 Avantor Performance
836006
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

57-50-1
C12H22O11
Sucrose
Crystalline
342.3
ACS Grade

Specifications

342.3
Crystalline
Poly
Sucrose
2.5kg
57-50-1
C12H22O11
ACS Grade