Please login to your online account to display your discounted pricing

Alfa Aesar™ Sucrose, ACS

$48.62 - $96.82

Chemical Identifiers

Chemical Name or Material Sucrose
CAS 57-50-1
Name Note For analysis ACS
Purity Grade ACS Grade
Molecular Formula C12H22O11
Formula Weight 342.3
MDL Number MFCD00006626
Beilstein 90825
Merck Index 14,8881
Synonym D-(+)-Saccharose
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA36508A1

 
alfa aesar™
36508A1
1kg Each for $96.82

AA3650830

 
alfa aesar
3650830*
250g Each for $48.62
Description & Specifications

Specifications

Formula Weight 342.3
Name Note For analysis ACS
Molecular Formula C12H22O11
Chemical Name or Material Sucrose
TSCA TSCA
Density 1.58g/mL
Purity Grade ACS Grade
Physical Form Crystalline
MDL Number MFCD00006626
CAS 57-50-1
Beilstein 90825
Merck Index 14,8881
Synonym D-(+)-Saccharose
Solubility Information Soluble in water; Slightly soluble in alcohol, methanol
Color White
Melting Point 185° to 187°C
Odor Odorless
RTECS Number WN6500000
Quantity 1kg