Please login to your online account to display your discounted pricing

Succinimide, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  348650050

 View more versions of this product

Catalog No. AC348650050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 99.08
Physical Form Powder or Flakes
Quantity 5g
Color Off-white to beige to light brown
RTECS Number WN2200000
EINECS Number 204-635-6
TSCA TSCA
Melting Point 121° to 126°C
Packaging Glass bottle
Synonym 2, 5-Pyrrolidinedione
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Succinimide
Name Note 98%
MDL Number MFCD00005495
Molecular Formula C4H5NO2
CAS 123-56-8
Assay Percent Range 98%

Specifications

Chemical Name or Material Succinimide
Name Note 98%
CAS 123-56-8
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H5NO2
Formula Weight 99.08
MDL Number MFCD00005495
Synonym 2, 5-Pyrrolidinedione
Accessories

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents